ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) 95%

Supplier: Ambeed
Warning

Synonyms: ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt

A626947-1G A626947-1MG A626947-250MG A626947-25G A626947-5G
76696-768EA 23 USD
76696-768 76696-770 76696-772 76696-774 77594-922
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) 95%
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])

Precaución: for Research Use Only

Formula: C₁₈H₂₄N₆O₆S₄
MW: 548.68 g/mol
MDL Number: MFCD00010404
CAS Number: 30931-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR