N-Benzyl-2-hydroxy-N,N-dimethylethanaminium chloride 98%

Supplier: Ambeed
Warning

A626765-100G A626765-1G A626765-25G A626765-5G
76696-760EA 477.41 USD
76696-760 76696-762 76696-764 76696-766
N-Benzyl-2-hydroxy-N,N-dimethylethanaminium chloride 98%
N-Benzyl-2-hydroxy-N,N-dimethylethanaminium chloride

Precaución: for Research Use Only

Formula: C₁₁H₁₈ClNO
MW: 215.71 g/mol
MDL Number: MFCD00040547
CAS Number: 7221-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR