7H-Benzo[c]fluoren-7-one 98%

Supplier: Ambeed
Warning

Synonyms: allo-Chrysoketone

A626111-1G A626111-250MG
76696-756EA 59.42 USD
76696-756 76696-758
7H-Benzo[c]fluoren-7-one 98%
7H-Benzo[c]fluoren-7-one

Precaución: for Research Use Only

Formula: C₁₇H₁₀O
MW: 230.27 g/mol
MDL Number: MFCD00215961
CAS Number: 6051-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR