N-Benzyl-4-methylaniline 97%

Supplier: Ambeed
Warning

A624120-100MG A624120-1G A624120-250MG A624120-5G
76696-748EA 63 USD
76696-748 76696-750 76696-752 76696-754
N-Benzyl-4-methylaniline 97%
N-Benzyl-4-methylaniline

Precaución: for Research Use Only

Formula: C₁₄H₁₅N
MDL Number: MFCD00870620
CAS Number: 5405-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR