Methyl-2,6-dimethoxybenzoate 95%

Supplier: Ambeed
Warning

A622618-10G A622618-1G A622618-250MG
76696-728EA 83 USD
76696-728 76696-730 76696-732
Methyl-2,6-dimethoxybenzoate 95%
Methyl-2,6-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MDL Number: MFCD00075861
CAS Number: 2065-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR