tri-Calcium phosphate 98%

Supplier: Ambeed
Warning

Synonyms: Calcium phosphate, Tricalcium orthophosphate, penta-Calcium hydroxide triphosphate, Calcium phosphate tribasic

A621074-1000G A621074-100G A621074-500G
76696-708EA 12 USD
76696-708 76696-710 76696-712
tri-Calcium phosphate 98%
tri-Calcium phosphate

Precaución: for Research Use Only

Formula: Ca₃(PO₄)₂
MW: 310.18 g/mol
MDL Number: MFCD00015984
CAS Number: 7758-87-4
Merck Index: 12,01741

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR