7,8-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one 97%

Supplier: Ambeed
Warning

Synonyms: YN1

A620652-250MG A620652-5G
76696-704EA 23.34 USD
76696-704 76696-706
7,8-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one 97%
7,8-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₅
MW: 270.237 g/mol
MDL Number: MFCD00885567
CAS Number: 75187-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR