(Chloromethyl)isopropoxydimethylsilane 96%

Supplier: Ambeed
Danger

Synonyms: Chloromethyl-dimethyl-propan-2-yloxysilane

A619478-1G A619478-5G A619478-25G
76696-686EA 11.67 USD
76696-686 76696-688 77602-564
(Chloromethyl)isopropoxydimethylsilane 96%
(Chloromethyl)isopropoxydimethylsilane

Precaución: for Research Use Only

Formula: C₆H₁₅ClOSi
MW: 166.72 g/mol
MDL Number: MFCD00010605
CAS Number: 18171-11-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR