Cyanomethylenetributylphosphorane 98%

Supplier: Ambeed
Warning

A615601-100G A615601-100MG A615601-10G A615601-1G A615601-250MG A615601-25G A615601-5G
76696-654EA 499 USD
76696-654 76696-656 76696-658 76696-660 76696-662 76696-664 76696-666
Cyanomethylenetributylphosphorane 98%
Cyanomethylenetributylphosphorane

Precaución: for Research Use Only

Formula: C₁₄H₂₈NP
MDL Number: MFCD01321151
CAS Number: 157141-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR