Benzophenone dimethyl acetal 98% (by GC)

Supplier: Ambeed
Warning

Synonyms: Benzophenone dimethyl ketal, Dimethoxydiphenylmethane

A614962-1G A614962-5G
76696-652EA 17 USD
76696-652 77878-427
Benzophenone dimethyl acetal 98% (by GC)
Benzophenone dimethyl acetal

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₂
MDL Number: MFCD00025900
CAS Number: 2235-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR