7-Methylimidazo[1,2-a]pyridine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A612177-100MG A612177-1G A612177-250MG A612177-50MG
76696-636EA 63.66 USD
76696-636 76696-638 76696-640 76696-642
7-Methylimidazo[1,2-a]pyridine-3-carboxylic acid 97%
7-Methylimidazo[1,2-a]pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
MDL Number: MFCD01720461
CAS Number: 21801-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR