ɑ-Pyrone 97%

Supplier: Ambeed
Warning

Synonyms: 2H-Pyran-2-one, alpha-Pyrone

A612059-100MG A612059-1G A612059-250MG A612059-5G A612059-25G
76696-628EA 28 USD
76696-628 76696-630 76696-632 77572-996 77878-423
ɑ-Pyrone 97%
alpha-Pyrone

Precaución: for Research Use Only

Formula: C₅H₄O₂
MDL Number: MFCD00006639
CAS Number: 504-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR