12-((tert-Butoxycarbonyl)amino)dodecanoic acid 98%

Supplier: Ambeed
Warning

A609740-1G A609740-250MG A609740-25G A609740-5G
76696-610EA 43.5 USD
76696-610 76696-612 76696-614 76696-616
12-((tert-Butoxycarbonyl)amino)dodecanoic acid 98%
12-((tert-Butoxycarbonyl)amino)dodecanoic acid

Precaución: for Research Use Only

Formula: C₁₇H₃₃NO₄
MW: 315.44 g/mol
MDL Number: MFCD00235887
CAS Number: 18934-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR