5-Benzyl-5-azaspiro[2.4]heptan-7-amine 98%

Supplier: Ambeed
Warning

A608520-250MG A608520-5G A608520-1G
76696-596EA 29 USD
76696-596 76696-598 77627-238
5-Benzyl-5-azaspiro[2.4]heptan-7-amine 98%
5-Benzyl-5-azaspiro[2.4]heptan-7-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂
MDL Number: MFCD09607966
CAS Number: 129306-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR