cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1R,2S)-2-(Boc-amino)cyclopentanecarboxylic Acid, cis-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid
A608170-100MG
A608170-1G
A608170-250MG
A608170-5G
76696-588EA
11
USD
76696-588
76696-590
76696-592
76696-594
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid 98%
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₄ MW: 229.276 g/mol |
MDL Number:
MFCD00800539 CAS Number: 136315-70-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...