Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3,5-dimethyl-2-pyrrolecarboxylate

A607912-100G A607912-10G A607912-1G A607912-25G A607912-5G
76696-578EA 396.78 USD
76696-578 76696-580 76696-582 76696-584 76696-586
Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate 98%
Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Number: MFCD00010645
CAS Number: 2199-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR