7,8-Dihydroisoquinolin-5(6H)-one 98%

Supplier: Ambeed
Warning

A607597-1G A607597-5G A607597-100MG
76696-576EA 231.28 USD
76696-576 77572-980 77594-856
7,8-Dihydroisoquinolin-5(6H)-one 98%
7,8-Dihydroisoquinolin-5(6H)-one

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.17 g/mol
MDL Number: MFCD08235026
CAS Number: 21917-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR