Ammonium tetraphenylborate 98%

Supplier: Ambeed
Warning

A604878-5G A604878-1G
76696-560EA 32 USD
76696-560 77878-406
Ammonium tetraphenylborate 98%
Ammonium tetraphenylborate

Precaución: for Research Use Only

Formula: C₂₄H₂₄BN
MDL Number: MFCD00134523
CAS Number: 14637-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR