4-(3,5-Bis(trimethylsilyl)benzamido)benzoic acid 95%

Supplier: Ambeed
Warning

A603641-100MG A603641-250MG A603641-50MG A603641-1G
76696-544EA 121 USD
76696-544 76696-546 76696-548 77878-402
4-(3,5-Bis(trimethylsilyl)benzamido)benzoic acid 95%
4-(3,5-Bis(trimethylsilyl)benzamido)benzoic acid

Precaución: for Research Use Only

Formula: C₂₀H₂₇NO₃Si₂
MDL Number: MFCD00952173
CAS Number: 125973-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR