1-(2,4-Dimethylphenyl)-2-thiourea 95%

Supplier: Ambeed
Danger

A603307-1G A603307-25G A603307-5G
76696-538EA 7 USD
76696-538 76696-540 76696-542
1-(2,4-Dimethylphenyl)-2-thiourea 95%
1-(2,4-Dimethylphenyl)-2-thiourea

Precaución: for Research Use Only

Formula: (C₆H₃(CH₃)₂)NC(S)NH₂
MDL Number: MFCD00041164
CAS Number: 16738-20-8
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR