Ammonium iron (II) sulfate hexahydrate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate
A602790-1000G
A602790-100G
A602790-25G
A602790-500G
76696-530EA
16.98
USD
76696-530
76696-532
76696-534
76696-536
Ammonium iron (II) sulfate hexahydrate 99%
Ammonium iron(II) sulphate hexahydrate
Precaución: for Research Use Only
Formula:
(NH₄)₂Fe(SO₄)₂·6H₂O MW: 392.14 g/mol |
MDL Number:
MFCD00150530 CAS Number: 7783-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...