3-Thiopheneboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: Thiophene-3-boronic acid pinacol ester, Thiophene-3-boronic acid, pinacol ester

A600995-10G A600995-1G A600995-250MG A600995-5G
76696-512EA 49 USD
76696-512 76696-514 76696-516 76696-518
3-Thiopheneboronic acid pinacol ester 96%
3-Thiopheneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₂S
MDL Number: MFCD05663893
CAS Number: 214360-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR