(R)-(-)-Mandelic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid
A600462-1000G
A600462-100G
A600462-10G
A600462-500G
A600462-25G
76696-504EA
70.02
USD
76696-504
76696-506
76696-508
76696-510
76844-264
(R)-(-)-Mandelic acid 97%
(R)-(-)-Mandelic acid
Precaución: for Research Use Only
Formula:
C₈H₈O₃ MW: 152.15 g/mol |
MDL Number:
MFCD00064251 CAS Number: 611-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...