Ethyl-7-(diethylamino)coumarin-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 7-(Diethylamino)coumarin-3-carboxylic Acid Ethyl Ester, Ethyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate

A599001-1G A599001-250MG A599001-5G
76696-498EA 45.62 USD
76696-498 76696-500 76696-502
Ethyl-7-(diethylamino)coumarin-3-carboxylate 98%
Ethyl-7-(diethylamino)coumarin-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₉NO₄
MW: 289.33 g/mol
MDL Number: MFCD00227484
CAS Number: 28705-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR