N-Benzyl-N-(3-isobutoxy-2-(pyrrolidin-1-yl)propyl)aniline hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Bepridil HCl

A598909-10MG A598909-1MG A598909-5MG
76696-492EA 106.09 USD
76696-492 76696-494 76696-496
N-Benzyl-N-(3-isobutoxy-2-(pyrrolidin-1-yl)propyl)aniline hydrochloride 97%
N-Benzyl-N-(3-isobutoxy-2-(pyrrolidin-1-yl)propyl)aniline hydrochloride

Precaución: for Research Use Only

Formula: C₂₄H₃₅ClN₂O
MW: 403 g/mol
MDL Number: MFCD00065498
CAS Number: 68099-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR