6-Nitroquinoxaline-2,3-dione 98%

Supplier: Ambeed
Warning

A597645-10G A597645-1G A597645-5G
76696-478EA 186.72 USD
76696-478 76696-480 76696-482
6-Nitroquinoxaline-2,3-dione 98%
6-Nitroquinoxaline-2,3-dione

Precaución: for Research Use Only

Formula: C₈H₃N₃O₄
MW: 205.13 g/mol
MDL Number: MFCD00673833
CAS Number: 1076197-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR