6-Phenoxynicotinic acid 98%

Supplier: Ambeed
Warning

A596743-1G A596743-5G
76696-474EA 64.72 USD
76696-474 76696-476
6-Phenoxynicotinic acid 98%
6-Phenoxynicotinic acid

Precaución: for Research Use Only

Formula: C₁₂H₉NO₃
MW: 215.2048 g/mol
MDL Number: MFCD02682068
CAS Number: 51362-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR