(9H-Fluoren-9-yl)methyl 3-aminopiperidine-1-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A592379-100MG A592379-1G
76696-458EA 72 USD
76696-458 76696-460
(9H-Fluoren-9-yl)methyl 3-aminopiperidine-1-carboxylate hydrochloride 97%
(9H-Fluoren-9-yl)methyl 3-aminopiperidine-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₂₃ClN₂O₂
MW: 358.86 g/mol
MDL Number: MFCD03001674
CAS Number: 811841-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR