4-(1-Pyrrolidinyl)piperidine 98%

Supplier: Ambeed
Danger

A591302-1G A591302-5G A591302-250MG
76696-454EA 19.1 USD
76696-454 76696-456 77627-224
4-(1-Pyrrolidinyl)piperidine 98%
4-(1-Pyrrolidinyl)piperidine

Precaución: for Research Use Only

Formula: C₉H₁₈N₂
MW: 154.2526 g/mol
MDL Number: MFCD00038011
CAS Number: 5004-07-9
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR