Bis(pentafluorophenyl)phenylphosphine 98%
Supplier: Ambeed
|
Warning
|
A590754-100MG
A590754-250MG
A590754-5G
76696-450EA
6
USD
76696-450
76696-452
77602-466
Bis(pentafluorophenyl)phenylphosphine 98%
Bis(pentafluorophenyl)phenylphosphine
Precaución: for Research Use Only
Formula:
C₁₈H₅F₁₀P |
MDL Number:
MFCD00000291 CAS Number: 5074-71-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...