Calpeptin 98%

Supplier: Ambeed
Warning

Synonyms: benzyl N-[(2S)-4-methyl-1-oxo-1-[[(2S)-1-oxohexan-2-yl]amino]pentan-2-yl]carbamate, Cbz-Leu-Nle-al, Benzyl ((S)-4-methyl-1-oxo-1-(((S)-1-oxohexan-2-yl)amino)pentan-2-yl)carbamate

A590110-10MG A590110-1MG A590110-25MG
76696-444EA 177 USD
76696-444 76696-446 76696-448
Calpeptin 98%
Calpeptin

Precaución: for Research Use Only

Formula: C₂₀H₃₀N₂O₄
MW: 362.47 g/mol
MDL Number: MFCD00155623
CAS Number: 117591-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR