3-Bromo-4-fluorocinnamic acid 98%

Supplier: Ambeed
Warning

A579591-1G A579591-25G A579591-5G
76696-402EA 8.49 USD
76696-402 76696-404 76696-406
3-Bromo-4-fluorocinnamic acid 98%
3-Bromo-4-fluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₆BrFO₂
MW: 245.05 g/mol
MDL Number: MFCD00143095
CAS Number: 160434-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR