Ethyl-2-nitrobenzoate 95%

Supplier: Ambeed
Warning

A578477-100G A578477-25G A578477-5G
76696-396EA 79 USD
76696-396 76696-398 76696-400
Ethyl-2-nitrobenzoate 95%
Ethyl-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MDL Number: MFCD00014696
CAS Number: 610-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR