cis-1,4-Diacetoxy-2-butene 98%

Supplier: Ambeed
Warning

Synonyms: cis-2-Butene-1,4-diol Diacetate, (Z)-But-2-ene-1,4-diyl diacetate

A577832-100G A577832-1G A577832-25G A577832-5G
76696-388EA 481.65 USD
76696-388 76696-390 76696-392 76696-394
cis-1,4-Diacetoxy-2-butene 98%
cis-1,4-Diacetoxy-2-butene

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
MDL Number: MFCD00059339
CAS Number: 25260-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR