(S)-1-(2, 4-Difluorophenyl)ethylamine 97%

Supplier: Ambeed
Danger

Synonyms: (1S)-1-(2,4-Difluorophenyl)ethanamine, (S)-1-(2,4-Difluorophenyl)ethanamine

A577534-100MG A577534-1G
76696-376EA 27.59 USD
76696-376 76696-378
(S)-1-(2, 4-Difluorophenyl)ethylamine 97%
(S)-1-(2, 4-Difluorophenyl)ethylamine

Precaución: for Research Use Only

Formula: C₈H₉F₂N
MW: 157.161 g/mol
MDL Number: MFCD06761834
CAS Number: 845252-02-0
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR