δ-Tridecanolactone 95%

Supplier: Ambeed
Warning

Synonyms: 6-Octyltetrahydro-2H-pyran-2-one, δ-Tridecalactone, Tetrahydro-6-n-octyl-2H-pyran-2-one

A576525-1G A576525-25G A576525-5G
76696-370EA 7.43 USD
76696-370 76696-372 76696-374
δ-Tridecanolactone 95%
delta-Tridecanolactone

Precaución: for Research Use Only

Formula: C₁₃H₂₄O₂
MW: 212.33 g/mol
MDL Number: MFCD00083590
CAS Number: 7370-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR