CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid) 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(Cyclohexylamino)-2-hydroxy-1-propanesulphonic acid
A575722-1000G
A575722-100G
A575722-10G
A575722-500G
A575722-25G
76696-354EA
137
USD
76696-354
76696-356
76696-358
76696-360
76844-124
CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid) 99%
CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid)
Precaución: for Research Use Only
Formula:
C₉H₁₉NO₄S |
MDL Number:
MFCD00041778 CAS Number: 73463-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...