Clorsulon 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-6-(1,2,2-trichlorovinyl)benzene-1,3-disulphonamide, 4-Amino-6-(trichlorovinyl)-1,3-benzenedisulphonamide

A574276-100MG A574276-25MG A574276-1G
76696-340EA 8.49 USD
76696-340 76696-342 77627-186
Clorsulon 98%
Clorsulon

Precaución: for Research Use Only

Formula: C₈H₈Cl₃N₃O₄S₂
MW: 380.66 g/mol
MDL Number: MFCD00072006
CAS Number: 60200-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR