6-Nitroquinoxaline 97%

Supplier: Ambeed
Warning

A573222-100MG A573222-10G A573222-1G A573222-250MG A573222-25G A573222-5G
76696-328EA 5 USD
76696-328 76696-330 76696-332 76696-334 76696-336 76696-338
6-Nitroquinoxaline 97%
6-Nitroquinoxaline

Precaución: for Research Use Only

Formula: C₈H₅N₃O₂
MW: 175.15
MDL Number: MFCD00462822
CAS Number: 6639-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR