Tris(triphenylphosphine)rhodium(I) bromide 98%

Supplier: Ambeed
Warning

Synonyms: Bromotris(triphenylphosphine)rhodium(I)

A572978-100MG A572978-250MG A572978-50MG
76696-322EA 85.94 USD
76696-322 76696-324 76696-326
Tris(triphenylphosphine)rhodium(I) bromide 98%
Tris(triphenylphosphine)rhodium(I) bromide

Precaución: for Research Use Only

Formula: C₅₄H₄₅BrP₃Rh
MW: 969.68 g/mol
MDL Number: MFCD00009828
CAS Number: 14973-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR