Cinnamonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3-Phenylacrylonitrile, Cinnamonitrile

A572452-25G A572452-500G A572452-5G
76696-318EA 10.61 USD
76696-318 76696-320 77627-182
Cinnamonitrile 97%
Cinnamonitrile

Precaución: for Research Use Only

Formula: C₉H₇N
MW: 129.16 g/mol
MDL Number: MFCD00001930
CAS Number: 4360-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR