1-Pyrenebutyric acid 95%

Supplier: Ambeed
Warning

A571530-100G A571530-10G A571530-1G A571530-25G A571530-5G
76696-308EA 1044.99 USD
76696-308 76696-310 76696-312 76696-314 76696-316
1-Pyrenebutyric acid 95%
1-Pyrenebutyric acid

Precaución: for Research Use Only

Formula: C₂₀H₁₆O₂
MW: 288.35 g/mol
MDL Number: MFCD00004141
CAS Number: 3443-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR