N-Benzoyl-D-alanine 98%

Supplier: Ambeed
Warning

Synonyms: Bz-D-Ala-OH

A571259-100G A571259-25G A571259-5G
76696-302EA 146 USD
76696-302 76696-304 76696-306
N-Benzoyl-D-alanine 98%
N-Benzoyl-D-alanine

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₃
MDL Number: MFCD00063127
CAS Number: 17966-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR