Potassium benzoate 99%

Supplier: Ambeed
Warning

Synonyms: Benzoic acid potassium salt

A570326-1000G A570326-100G A570326-25G A570326-500G
76696-294EA 31 USD
76696-294 76696-296 76696-298 76696-300
Potassium benzoate 99%
Potassium benzoate

Precaución: for Research Use Only

Formula: C₇H₅KO₂
MW: 160.21 g/mol
MDL Number: MFCD00013061
CAS Number: 582-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR