2-Amino-5-isopropylbenzonitrile 97%

Supplier: Ambeed
Warning

A569268-1G A569268-250MG
76696-274EA 145 USD
76696-274 76696-276
2-Amino-5-isopropylbenzonitrile 97%
2-Amino-5-isopropylbenzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂
MDL Number: MFCD11977495
CAS Number: 549488-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR