6-Undecanol 95%

Supplier: Ambeed
Danger

Synonyms: 6-Undecyl alcohol, 6-Hydroxy undecane

A569105-1G A569105-25G
76696-272EA 9.55 USD
76696-272 77572-810
6-Undecanol 95%
6-Undecanol

Precaución: for Research Use Only

Formula: C₁₁H₂₄O
MW: 172.31 g/mol
MDL Number: MFCD00021951
CAS Number: 23708-56-7
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR