2,5-Dioxopyrrolidin-1-yl 6-acrylamidohexanoate 97%

Supplier: Ambeed
Warning

A566633-100MG A566633-250MG
76696-256EA 131.56 USD
76696-256 76696-258
2,5-Dioxopyrrolidin-1-yl 6-acrylamidohexanoate 97%
2,5-Dioxopyrrolidin-1-yl 6-acrylamidohexanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₅
MW: 282.29 g/mol
MDL Number: MFCD03427755
CAS Number: 63392-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR