4,5-Dimethoxy-2-nitrobenzyl carbonochloridate 97%

Supplier: Ambeed
Danger

A562083-100MG A562083-250MG A562083-1G A562083-5G A562083-25G
76696-226EA 15 USD
76696-226 76696-228 77551-688 77572-780 77878-357
4,5-Dimethoxy-2-nitrobenzyl carbonochloridate 97%
4,5-Dimethoxy-2-nitrobenzyl carbonochloridate

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClNO₆
MDL Number: MFCD00143507
CAS Number: 42855-00-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR