7-Methoxycoumarin-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A559072-100MG A559072-1G A559072-250MG A559072-25G A559072-5G
76696-212EA 7 USD
76696-212 76696-214 76696-216 77627-150 77627-152
7-Methoxycoumarin-3-carboxylic acid 98%
7-Methoxycoumarin-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₈O₅
MDL Number: MFCD00452770
CAS Number: 20300-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR