2-Propoxybenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-n-Propoxybenzaldehyde

A558783-1G A558783-250MG A558783-5G
76696-204EA 42.44 USD
76696-204 76696-206 76696-208
2-Propoxybenzaldehyde 98%
2-Propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00046150
CAS Number: 7091-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR